Assay Details
| Gene Symbol: LOC4341387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788295.2 | XP_015643781.1 | 5 - 6 | 1468 | 102 | |
| GenBank mRNA | AF513382.1 | - | 5 - 6 | 1433 | 102 | |
| AK068682.1 | - | 5 - 6 | 1450 | 102 | ||
| BT131785.1 | - | 2 - 3 | 147 | 102 | ||
More Information
Set Membership: |
|

LOC4341387
Genomic Map