Assay Details
| Gene Symbol: LOC4341387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788295.2 | XP_015643781.1 | 6 - 7 | 1558 | 72 | |
| GenBank mRNA | AF513382.1 | - | 6 - 7 | 1523 | 72 | |
| AK068682.1 | - | 6 - 7 | 1540 | 72 | ||
| BT131785.1 | - | 3 - 4 | 237 | 72 | ||
More Information
Set Membership: |
|

LOC4341387
Genomic Map