Assay Details
| Gene Symbol: LOC9266513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789954.2 | XP_015645440.1 | 14 - 15 | 1396 | 67 | |
| GenBank mRNA | AK069343.1 | - | 11 - 12 | 2393 | 67 | |
More Information
Set Membership: |
|

LOC9266513
Genomic Map