Assay Details
| Gene Symbol: LOC9266513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789954.2 | XP_015645440.1 | 23 - 24 | 2277 | 63 | |
| GenBank mRNA | AK069343.1 | - | 19 - 20 | 3364 | 63 | |
More Information
Set Membership: |
|

LOC9266513
Genomic Map