Assay Details
| Gene Symbol: LOC4343899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792050.1 | XP_015647536.1 | 2 - 3 | 539 | 88 | |
| GenBank mRNA | AK065604.1 | - | 2 - 3 | 471 | 88 | |
| AK068566.1 | - | 2 - 3 | 469 | 88 | ||
More Information
Set Membership: |
|

LOC4343899
Genomic Map