Assay Details
| Gene Symbol: LOC4344425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795388.2 | XP_015650874.1 | 1 | 1144 | 137 | |
| GenBank mRNA | AK061430.1 | - | 1 | 806 | 137 | |
| AK061546.1 | - | 1 | 713 | 137 | ||
| AK061780.1 | - | 1 | 825 | 137 | ||
| AK067526.1 | - | 1 | 1048 | 137 | ||
| AK072003.1 | - | 1 | 947 | 137 | ||
| AK099239.1 | - | 1 | 970 | 137 | ||
| AK104488.1 | - | 1 | 802 | 137 | ||
More Information
Set Membership: |
|

LOC4344425
Genomic Map