Assay Details
| Gene Symbol: LOC4345402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792839.2 | XP_015648325.1 | 11 - 12 | 967 | 102 | |
| XM_015792840.2 | XP_015648326.1 | 11 - 12 | 880 | 102 | ||
More Information
Set Membership: |
|

LOC4345402
Genomic Map