Assay Details
| Gene Symbol: LOC4345402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792839.2 | XP_015648325.1 | 3 - 4 | 369 | 113 | |
| XM_015792840.2 | XP_015648326.1 | 3 - 4 | 282 | 113 | ||
More Information
Set Membership: |
|

LOC4345402
Genomic Map