Assay Details
| Gene Symbol: LOC4325471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765488.1 | XP_015620974.1 | 2 - 3 | 781 | 87 | |
| GenBank mRNA | AK058378.1 | - | 2 - 3 | 706 | 87 | |
More Information
Set Membership: |
|

LOC4325471
Genomic Map