Assay Details
| Gene Symbol: LOC4325879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766159.2 | XP_015621645.1 | 5 - 6 | 687 | 79 | |
| GenBank mRNA | AK071126.1 | - | 4 - 5 | 707 | 79 | |
More Information
Set Membership: |
|

LOC4325879
Genomic Map