Assay Details
| Gene Symbol: LOC4325879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766159.2 | XP_015621645.1 | 7 - 8 | 869 | 80 | |
| GenBank mRNA | AK071126.1 | - | 6 - 7 | 889 | 80 | |
More Information
Set Membership: |
|

LOC4325879
Genomic Map