Assay Details
| Gene Symbol: LOC4324191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766133.2 | XP_015621619.1 | 8 - 9 | 1039 | 67 | |
| GenBank mRNA | AK103045.1 | - | 8 - 9 | 990 | 67 | |
More Information
Set Membership: |
|

LOC4324191
Genomic Map