Assay Details
| Gene Symbol: LOC4324985 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758960.2 | XP_015614446.1 | 2 - 3 | 413 | 64 | |
| GenBank mRNA | AK067269.1 | - | 2 - 3 | 394 | 64 | |
More Information
Set Membership: |
|

LOC4324985
Genomic Map