Assay Details
| Gene Symbol: LOC4325388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776298.2 | XP_015631784.1 | 1 | 1283 | 142 | |
| GenBank mRNA | AK107370.1 | - | 1 | 1152 | 142 | |
More Information
Set Membership: |
|

LOC4325388
Genomic Map