Assay Details
| Gene Symbol: LOC4348060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759009.2 | XP_015614495.1 | 2 - 3 | 505 | 69 | |
| GenBank mRNA | AK066939.1 | - | 2 - 3 | 490 | 69 | |
| AK098985.1 | - | 2 - 3 | 490 | 69 | ||
More Information
Set Membership: |
|

LOC4348060
Genomic Map