Assay Details
| Gene Symbol: LOC4349116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757235.2 | XP_015612721.1 | 15 - 16 | 1600 | 95 | |
| GenBank mRNA | AK065934.1 | - | 16 - 17 | 1402 | 95 | |
More Information
Set Membership: |
|

LOC4349116
Genomic Map