Assay Details
| Gene Symbol: LOC4349475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758696.2 | XP_015614182.1 | 2 - 3 | 558 | 156 | |
| XM_015758697.2 | XP_015614183.1 | 2 - 3 | 616 | 156 | ||
| GenBank mRNA | AK099409.1 | - | 2 - 3 | 514 | 156 | |
| AK103988.1 | - | 2 - 3 | 495 | 156 | ||
More Information
Set Membership: |
|

LOC4349475
Genomic Map