Assay Details
| Gene Symbol: LOC4350526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759702.2 | XP_015615188.1 | 1 - 2 | 1099 | 69 | |
| GenBank mRNA | AK121422.1 | - | 1 - 2 | 88 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015759702.2 transcripts.
Set Membership: |
|

LOC4350526
Genomic Map