Assay Details
| Gene Symbol: LOC4350526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759702.2 | XP_015615188.1 | 4 - 5 | 1727 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4350526| Gene Symbol: LOC4350526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759702.2 | XP_015615188.1 | 4 - 5 | 1727 | 112 | |
Set Membership: |
|