Assay Details
| Gene Symbol: LOC4350526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759702.2 | XP_015615188.1 | 5 - 6 | 1922 | 138 | |
| GenBank mRNA | AK121422.1 | - | 5 - 6 | 911 | 138 | |
More Information
Set Membership: |
|

LOC4350526
Genomic Map