Assay Details
| Gene Symbol: LOC4350526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759702.2 | XP_015615188.1 | 6 - 7 | 2139 | 85 | |
| GenBank mRNA | AK121422.1 | - | 6 - 7 | 1128 | 85 | |
More Information
Set Membership: |
|

LOC4350526
Genomic Map