Assay Details
| Gene Symbol: LOC4327920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777552.2 | XP_015633038.1 | 1 - 2 | 261 | 65 | |
| GenBank mRNA | AK072598.1 | - | 1 - 2 | 265 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015777552.2 transcripts.
Set Membership: |
|

LOC4327920
Genomic Map