Assay Details
| Gene Symbol: LOC4327920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777524.2 | XP_015633010.1 | 5 - 6 | 849 | 71 | |
| XM_015777533.2 | XP_015633019.1 | 5 - 6 | 666 | 71 | ||
| XM_015777542.2 | XP_015633028.1 | 5 - 6 | 849 | 71 | ||
| XM_015777552.2 | XP_015633038.1 | 5 - 6 | 763 | 71 | ||
| GenBank mRNA | AK072598.1 | - | 5 - 6 | 767 | 71 | |
More Information
Set Membership: |
|

LOC4327920
Genomic Map