Assay Details
| Gene Symbol: LOC4327940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775312.2 | XP_015630798.1 | 1 - 2 | 766 | 120 | |
| GenBank mRNA | AK072744.1 | - | 1 - 2 | 709 | 120 | |
| AK099957.1 | - | 1 - 2 | 665 | 120 | ||
More Information
Set Membership: |
|

LOC4327940
Genomic Map