Assay Details
Gene Symbol: LOC4327960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770176.2 | XP_015625662.1 | 4 - 5 | 488 | 101 | |
GenBank mRNA | AK064082.1 | - | 4 - 5 | 378 | 101 | |
AK101426.1 | - | 4 - 5 | 382 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
