Assay Details
| Gene Symbol: LOC4328369 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770780.2 | XP_015626266.1 | 1 - 2 | 580 | 77 | |
| GenBank mRNA | AK108324.1 | - | 1 - 2 | 584 | 77 | |
More Information
Set Membership: |
|

LOC4328369
Genomic Map