Assay Details
| Gene Symbol: LOC4328992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771712.2 | XP_015627198.1 | 2 - 3 | 302 | 151 | |
| GenBank mRNA | AK099165.1 | - | 2 - 3 | 258 | 151 | |
More Information
Set Membership: |
|

LOC4328992
Genomic Map