Assay Details
| Gene Symbol: LOC4328992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771712.2 | XP_015627198.1 | 4 - 5 | 1157 | 74 | |
| GenBank mRNA | AK061985.1 | - | 4 - 5 | 1146 | 74 | |
| AK099165.1 | - | 4 - 5 | 1113 | 74 | ||
More Information
Set Membership: |
|

LOC4328992
Genomic Map