Assay Details
| Gene Symbol: LOC4329368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771480.2 | XP_015626966.1 | 16 - 17 | 1942 | 101 | |
| XM_015771485.2 | XP_015626971.1 | 16 - 17 | 1861 | 101 | ||
| GenBank mRNA | AK066344.1 | - | 16 - 17 | 1760 | 101 | |
More Information
Set Membership: |
|

LOC4329368
Genomic Map