Assay Details
| Gene Symbol: LOC4329368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771480.2 | XP_015626966.1 | 8 - 9 | 1163 | 149 | |
| XM_015771485.2 | XP_015626971.1 | 8 - 9 | 1094 | 149 | ||
| GenBank mRNA | AK066344.1 | - | 8 - 9 | 993 | 149 | |
More Information
Set Membership: |
|

LOC4329368
Genomic Map