Assay Details
| Gene Symbol: LOC4329578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771919.2 | XP_015627405.1 | 6 - 7 | 768 | 60 | |
| GenBank mRNA | AK068919.1 | - | 6 - 7 | 745 | 60 | |
| AK104703.1 | - | 6 - 7 | 744 | 60 | ||
| AM039889.1 | - | 6 - 7 | 673 | 60 | ||
| BT131430.1 | - | 1 - 2 | 78 | 60 | ||
More Information
Set Membership: |
|

LOC4329578
Genomic Map