Assay Details
| Gene Symbol: LOC4329911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770238.2 | XP_015625724.1 | 12 - 13 | 1530 | 103 | |
| GenBank mRNA | AK105762.1 | - | 12 - 13 | 1505 | 103 | |
| AK120769.1 | - | 12 - 13 | 1498 | 103 | ||
| AY224457.1 | - | 9 - 10 | 936 | 103 | ||
More Information
Set Membership: |
|

LOC4329911
Genomic Map