Assay Details
| Gene Symbol: LOC4329911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770238.2 | XP_015625724.1 | 16 - 17 | 1952 | 80 | |
| GenBank mRNA | AK105762.1 | - | 16 - 17 | 1927 | 80 | |
| AK120769.1 | - | 16 - 17 | 1920 | 80 | ||
| AY224457.1 | - | 13 - 14 | 1358 | 80 | ||
More Information
Set Membership: |
|

LOC4329911
Genomic Map