Assay Details
| Gene Symbol: LOC4330086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767559.2 | XP_015623045.1 | 2 - 3 | 407 | 65 | |
| GenBank mRNA | AK069779.1 | - | 2 - 3 | 353 | 65 | |
More Information
Set Membership: |
|

LOC4330086
Genomic Map