Assay Details
| Gene Symbol: LOC4330539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770159.2 | XP_015625645.1 | 1 - 2 | 162 | 116 | |
| GenBank mRNA | AK073448.1 | - | 1 - 2 | 147 | 116 | |
| AK100143.1 | - | 1 - 2 | 106 | 116 | ||
| EF122491.1 | - | 1 - 2 | 97 | 116 | ||
| GQ848044.1 | - | 1 - 2 | 94 | 116 | ||
| JF969212.1 | - | 1 - 2 | 92 | 116 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770159.2 transcripts.
Set Membership: |
|

LOC4330539
Genomic Map