Assay Details
| Gene Symbol: LOC4330539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770159.2 | XP_015625645.1 | 3 - 4 | 1032 | 97 | |
| GenBank mRNA | AK073448.1 | - | 3 - 4 | 1017 | 97 | |
| AK100143.1 | - | 3 - 4 | 976 | 97 | ||
| AK119207.1 | - | 3 - 4 | 977 | 97 | ||
| D12637.1 | - | 3 - 4 | 964 | 97 | ||
| EF122491.1 | - | 3 - 4 | 967 | 97 | ||
| EU605818.1 | - | 2 - 3 | 852 | 97 | ||
| GQ848044.1 | - | 3 - 4 | 964 | 97 | ||
| JF969211.1 | - | 3 - 4 | 943 | 97 | ||
| JF969212.1 | - | 3 - 4 | 962 | 97 | ||
More Information
Set Membership: |
|

LOC4330539
Genomic Map