Assay Details
| Gene Symbol: LOC4330573 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767674.2 | XP_015623160.1 | 4 - 5 | 427 | 96 | |
| XM_015767675.1 | XP_015623161.1 | 4 - 5 | 427 | 96 | ||
| GenBank mRNA | AK073256.1 | - | 3 - 4 | 280 | 96 | |
| AK105251.1 | - | 1235 | 96 | |||
More Information
Set Membership: |
|

LOC4330573
Genomic Map