Assay Details
| Gene Symbol: LOC4330852 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771029.2 | XP_015626515.1 | 1 - 2 | 518 | 157 | |
| GenBank mRNA | AK059725.1 | - | 1 - 2 | 478 | 157 | |
| AK060304.1 | - | 2 - 3 | 416 | 157 | ||
| AK098996.1 | - | 1 - 2 | 475 | 157 | ||
| AK099120.1 | - | 1 - 2 | 474 | 157 | ||
| AK104659.1 | - | 1 - 2 | 473 | 157 | ||
More Information
Set Membership: |
|

LOC4330852
Genomic Map