Assay Details
| Gene Symbol: LOC4330852 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771029.2 | XP_015626515.1 | 4 - 5 | 898 | 93 | |
| GenBank mRNA | AK059725.1 | - | 4 - 5 | 858 | 93 | |
| AK060304.1 | - | 5 - 6 | 796 | 93 | ||
| AK098996.1 | - | 4 - 5 | 855 | 93 | ||
| AK104659.1 | - | 4 - 5 | 853 | 93 | ||
More Information
Set Membership: |
|

LOC4330852
Genomic Map