Assay Details
| Gene Symbol: LOC4332110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773134.2 | XP_015628620.1 | 2 - 3 | 477 | 79 | |
| GenBank mRNA | AK066490.1 | - | 2 - 3 | 478 | 79 | |
More Information
Set Membership: |
|

LOC4332110
Genomic Map