Assay Details
| Gene Symbol: LOC4333399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776158.2 | XP_015631644.1 | 5 - 6 | 1234 | 88 | |
| GenBank mRNA | AB084385.1 | - | 5 - 6 | 1014 | 88 | |
| AK072295.1 | - | 5 - 6 | 1235 | 88 | ||
| AK101670.1 | - | 5 - 6 | 1235 | 88 | ||
More Information
Set Membership: |
|

LOC4333399
Genomic Map