Assay Details
| Gene Symbol: LOC4333399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776158.2 | XP_015631644.1 | 6 - 7 | 1311 | 115 | |
| GenBank mRNA | AB084385.1 | - | 6 - 7 | 1091 | 115 | |
| AK072295.1 | - | 6 - 7 | 1312 | 115 | ||
| AK101670.1 | - | 6 - 7 | 1312 | 115 | ||
More Information
Set Membership: |
|

LOC4333399
Genomic Map