Assay Details
| Gene Symbol: LOC4334387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775107.2 | XP_015630593.1 | 2 - 3 | 979 | 149 | |
| GenBank mRNA | AK064808.1 | - | 3 - 4 | 796 | 149 | |
More Information
Set Membership: |
|

LOC4334387
Genomic Map