Assay Details
| Gene Symbol: LOC4334893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777953.1 | XP_015633439.1 | 15 - 16 | 1798 | 153 | |
| XM_015777954.2 | XP_015633440.1 | 15 - 16 | 1798 | 153 | ||
| GenBank mRNA | AK101740.1 | - | 15 - 16 | 1665 | 153 | |
| DQ118178.1 | - | 15 - 16 | 1524 | 153 | ||
More Information
Set Membership: |
|

LOC4334893
Genomic Map