Assay Details
| Gene Symbol: LOC4334893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777953.1 | XP_015633439.1 | 18 - 19 | 2018 | 62 | |
| GenBank mRNA | AK101740.1 | - | 18 - 19 | 1885 | 62 | |
| DQ118178.1 | - | 18 - 19 | 1744 | 62 | ||
More Information
Set Membership: |
|

LOC4334893
Genomic Map