Assay Details
| Gene Symbol: LOC4335313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778447.2 | XP_015633933.1 | 6 - 7 | 839 | 80 | |
| GenBank mRNA | AK119369.1 | - | 5 - 6 | 1369 | 80 | |
More Information
Set Membership: |
|

LOC4335313
Genomic Map