Assay Details
| Gene Symbol: LOC4335313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778447.2 | XP_015633933.1 | 9 - 10 | 1196 | 122 | |
| GenBank mRNA | AK119369.1 | - | 8 - 9 | 1726 | 122 | |
More Information
Set Membership: |
|

LOC4335313
Genomic Map