Assay Details
| Gene Symbol: LOC4336755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780890.1 | XP_015636376.1 | 2 - 3 | 1001 | 89 | |
| GenBank mRNA | AK072398.1 | - | 2 - 3 | 1007 | 89 | |
More Information
Set Membership: |
|

LOC4336755
Genomic Map