Assay Details
| Gene Symbol: LOC4336755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780890.1 | XP_015636376.1 | 7 - 8 | 1827 | 82 | |
| GenBank mRNA | AK104058.1 | - | 2 - 3 | 193 | 82 | |
More Information
Set Membership: |
|

LOC4336755
Genomic Map