Assay Details
| Gene Symbol: LOC4337088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780662.2 | XP_015636148.1 | 10 - 11 | 1177 | 130 | |
| GenBank mRNA | AK065665.1 | - | 10 - 11 | 1184 | 130 | |
| AK099320.1 | - | 10 - 11 | 1134 | 130 | ||
More Information
Set Membership: |
|

LOC4337088
Genomic Map